Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclohexylzinc Bromide Solution

Catalog No. 49803350ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-803-350ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 49-803-350ML Supplier Sigma Aldrich Supplier No. 49803350ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cyclohexylzinc bromide is an organozinc reagent, generally used in Negishi coupling. Examples include the cross-coupling of alkylzinc halides with aryl/heteroaryl halides using Pd-PEPPSI-IPent and nickel-catalyzed synthesis of 4-substituted coumarins.It can react with the SO2 surrogate, 1,4-diazabicyclo[2.2.2]octane bis(sulfur dioxide) adduct (DABSO) to afford zinc sulfinate salts which can be alkylated to synthesize various useful sulfones.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 131379-39-0
Density 0.976 g/mL at 25°C
Molecular Formula C6H11BrO2Zn
Linear Formula C2H5O2C(CH2)3ZnBr
MDL Number MFCD00671977
Quantity 50 mL
Molecular Weight (g/mol) 260.44 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.