accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13138-53-9

IUPAC Name: 4-nitrobenzene-1,2-dicarboxamide
Synonyms: 4-nitrobenzene-1,2-dicarboxamide
Molecular Weight (g/mol): 209.16
Molecular Formula: C8H7N3O4
InChi Key: XWCDSCYRIROFIO-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(C=C1C(N)=O)[N+]([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results