Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophthalamide, TCI America™
SDP

Catalog No. N06475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06475G Supplier TCI America Supplier No. N06475G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13138-53-9
Molecular Formula C8H7N3O4
Molecular Weight (g/mol) 209.16
MDL Number MFCD00052702
InChI Key XWCDSCYRIROFIO-UHFFFAOYSA-N
Synonym 4-nitrophthalamide, 4-nitrophthaldiamide, 1,2-benzenedicarboxamide, 4-nitro, 1,2-benzenedicarboxamide,4-nitro, acmc-209bll, maybridge1_003271, 4-nitrophthalamide 1g, xwcdscyrirofio-uhfffaoysa
PubChem CID 83167
IUPAC Name 4-nitrobenzene-1,2-dicarboxamide
SMILES NC(=O)C1=CC=C(C=C1C(N)=O)[N+]([O-])=O

Specifications

Melting Point 195°C
Color Yellow
Quantity 5 g
Formula Weight 209.16
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophthalamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.