CAS RN 1313900-20-7
IUPAC Name:
2,7-dibromo-9-(4-bromophenyl)-9H-carbazole
Synonyms:
2,7-dibromo-9-(4-bromophenyl)carbazole
Molecular Weight (g/mol):
480
Molecular Formula:
C18H10Br3N
InChi Key:
MKSHSKZVLIHXMT-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)N1C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
1
–
1
of
1
results
1
2,7-Dibromo-9-(4-bromophenyl)-9H-carbazole 98.0+%, TCI America™
CAS: 1313900-20-7 Molecular Formula: C18H10Br3N Molecular Weight (g/mol): 479.997 InChI Key: MKSHSKZVLIHXMT-UHFFFAOYSA-N PubChem CID: 129317820 IUPAC Name: 2,7-dibromo-9-(4-bromophenyl)carbazole SMILES: C1=CC(=CC=C1N2C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br)Br