accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOH2OH2OH2OH2OH2OH2OMg

CAS RN 131391-55-4

IUPAC Name: magnesium(2+) 2-carboperoxoatobenzoate hexahydrate
Synonyms: magnesium(2+) 2-carboperoxoatobenzoate hexahydrate
Molecular Weight (g/mol): 312.51
Molecular Formula: C8H16MgO11
InChi Key: FODOUIXGKGNSMR-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.[Mg++].[O-]OC(=O)C1=CC=CC=C1C([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results