Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium monoperoxyphthalate hexahydrate, tech. ca 80%, Thermo Scientific™

Catalog No. AAB2056530
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2056530 250 g
AAB2056514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2056530 Supplier Thermo Scientific Chemicals Supplier No. B2056530

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131391-55-4
Molecular Formula C8H16MgO11
Molecular Weight (g/mol) 312.51
MDL Number MFCD00013096
InChI Key FODOUIXGKGNSMR-UHFFFAOYSA-L
Synonym magnesium monoperoxyphthalate hexahydrate, magnesium monoperoxyphthalate, mmpp, magnesium monoperoxyphtalate hexahydrate, magnesium monoperoxyphathalate hexahydrate, monoperoxy-phthalic acid magnesium salt hexahydrate, monoperoxyphthalic acid magnesium salt hexahydrate, magnesium 2+ 2-carboperoxoatobenzoate hexahydrate, magnesium 2+ ion 2-carboperoxoatobenzoate hexahydrate, magnesium 2-dioxidan-2-ide-1-carbonyl benzoate-water 1/1/6
PubChem CID 21894687
IUPAC Name magnesium;2-oxidooxycarbonylbenzoate;hexahydrate
SMILES C1=CC=C(C(=C1)C(=O)[O-])C(=O)O[O-].O.O.O.O.O.O.[Mg+2]

Specifications

Melting Point ∼93°C (decomposition)
Quantity 250 g
Merck Index 14,5673
Solubility Information Soluble in water
Formula Weight 494.65 (386.55 Anhydrous)
Percent Purity ∼80%
Grade Technical
Chemical Name or Material Magnesium monoperoxyphthalate hexahydrate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.