accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1314-34-7

IUPAC Name: oxo[(oxovanadio)oxy]vanadium
Synonyms: vanadium(III) oxide
Molecular Weight (g/mol): 149.88
Molecular Formula: O3V2
InChi Key: KFAFTZQGYMGWLU-UHFFFAOYSA-N
SMILES: O=[V]O[V]=O
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (6)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)