Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Vanadium(III) oxide, 99.7% (metals basis)

Catalog No. AA4337714
Change view
Click to view available options
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA4337714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4337714 Supplier Thermo Scientific™ Supplier No. 4337714
Add to Cart

Specifications

Melting Point 1970°
CAS 1314-34-7
Physical Form Solid
Quantity 25 g
Molecular Formula V{2}O{3}
MDL Number MFCD00011455
UN Number UN3285
Merck Index 14,9923
Synonym Vanadium trioxide;
Formula Weight 149.88
Percent Purity 99.7%
Density 4.87
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.