CAS RN 1314-36-9
Yttrium(III) oxide, 99.999%, (trace metal basis)
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium(III) oxide, 99.99%, (trace metal basis)
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium, plasma standard solution, Specpure™ Y 1000μg/mL
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium(III) oxide, REacton™, 99.99% (REO)
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium(III) oxide, REacton™, 99.9% (REO)
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium(III) oxide, REacton™, nanopowder, 99.995% (REO)
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium(III) oxide, REacton™, 99.9% (REO)
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N Synonym: Yttria IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium(III) oxide, nanopowder, 99.995% (REO)
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium(III) oxide, REacton™, 99.99% (REO)
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium(III) oxide, REacton™, 99.999% (REO)
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium(III) oxide pellets, 99.9% (REO), Thermo Scientific Chemicals
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium(III) oxide, REacton™, 99.9999% (REO)
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium Oxide, 99%+, MilliporeSigma™
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N Synonym: Yttrium (III) Oxide 99 IUPAC Name: Diyttrium(3+) Trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium ICP Standard, 1000 ppm Y in 3% HNO3, Ricca Chemical
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Yttrium AA Standard, 1000 ppm Y in 3% HNO3, Ricca Chemical
CAS: 1314-36-9 Molecular Formula: O3Y2 Molecular Weight (g/mol): 225.81 MDL Number: MFCD00011473 InChI Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N IUPAC Name: diyttrium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
- 1
- 2