Learn More
Yttrium(III) oxide pellets, 99.9% (REO), Thermo Scientific Chemicals
Description
Sintered pellets suitable for vacuum depositionYttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is a precursor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2410°C |
| Quantity | 100 g |
| Odor | Odorless |
| Sensitivity | Air Sensitive |
| Density | 5.01 g/mL |
| Merck Index | 14,10107 |
| Solubility Information | Insoluble in water. Soluble in alcohol and dilute acids. |
| Formula Weight | 225.81 |
| Percent Purity | 99.9% |
| Assay | (REO) |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 215-233-5 |
| RTECSNumber | ZG3850000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.