accessibility menu, dialog, popup

UserName

Viewing profile as user:
OVOOVOO

CAS RN 1314-62-1

IUPAC Name: [(dioxovanadio)oxy]dioxovanadium
Synonyms: vanadium(V) oxide|divanadium pentaoxide|vanadium pentoxide (orthorhombic crystalline form)
Molecular Weight (g/mol): 181.88
Molecular Formula: O5V2
InChi Key: GNTDGMZSJNCJKK-UHFFFAOYSA-N
SMILES: O=[V](=O)O[V](=O)=O
Molecular Weight (g/mol) 
  • (34)
  • (5)
  • (6)
Quantity 
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (6)
  • (3)
  • (6)
  • (2)
Grade 
  • (2)
  • (1)
  • (6)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (2)
  • (7)
  • (8)
  • (6)
  • (22)
  • (34)
  • (5)
  • (6)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (3)
  • (1)
  • (2)
  • (6)
  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (6)
  • (3)