Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadium(V) oxide, 99.99% (metals basis)

Catalog No. AA1090409
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1090409 10 g
AA1090404 2 g
AA1090418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1090409 Supplier Thermo Scientific Chemicals Supplier No. 01090409
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1314-62-1 | O5V2 | 181.88 g/mol

Vanadium pentoxide is the precursor to alloys of vanadium. It is widely used as an industrial catalyst for the manufacture of sulfuric acid, maleic anhydride, phthalic anhydride, adipic acid, acrylic acid, oxalic acid and anthraquinone. It finds application as a detector material in bolometers and microbolometer arrays for thermal imaging, and as an ethanol sensor in ppm (parts per million) levels. It is also used in vanadium redox batteries, which are used for energy storage in power facilities, such as wind farms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-62-1
Molecular Formula O5V2
Molecular Weight (g/mol) 181.88
MDL Number MFCD00011457
InChI Key GNTDGMZSJNCJKK-UHFFFAOYSA-N
Synonym vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
PubChem CID 14814
ChEBI CHEBI:30045
SMILES O=[V](=O)O[V](=O)=O

Specifications

Boiling Point 1750°C (decomposition)
Melting Point 690°C
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Slightly soluble in water.
IUPAC Name [(dioxovanadio)oxy]dioxovanadium
Formula Weight 181.88
Density 3.357 g/mL
Chemical Name or Material Vanadium(V) oxide
Hazard Category H302+H332-H318-H335-H341-H351-H361d-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P310-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 215-239-8
RTECSNumber YW2125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.