accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNO

CAS RN 1315281-09-4

IUPAC Name: 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)propyl]morpholine
Synonyms: 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)propyl]morpholine
Molecular Weight (g/mol): 255.17
Molecular Formula: C13H26BNO3
InChi Key: CBGALTGWZUGSAO-UHFFFAOYSA-N
SMILES: CC1(C)OB(CCCN2CCOCC2)OC1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results