Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Morpholinyl)propyl-1-boronic acid pinacol ester, 98%, Thermo Scientific™

Catalog No. AAH3019103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3019103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3019103 Supplier Thermo Scientific Chemicals Supplier No. H3019103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315281-09-4
Molecular Formula C13H26BNO3
Molecular Weight (g/mol) 255.165
MDL Number MFCD12546182
InChI Key CBGALTGWZUGSAO-UHFFFAOYSA-N
Synonym 3-4-morpholinyl propyl-1-boronic acid pinacol ester, 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl propyl morpholine
PubChem CID 73995111
IUPAC Name 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)propyl]morpholine
SMILES B1(OC(C(O1)(C)C)(C)C)CCCN2CCOCC2

Specifications

Refractive Index 1.4585
Quantity 1 g
Formula Weight 255.17
Percent Purity 98%
Chemical Name or Material 3-(4-Morpholinyl)propyl-1-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.