CAS RN 131570-56-4
CAS RN 131570-56-4
IUPAC Name:
2-{[(tert-butoxy)carbonyl]amino}-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms:
2-[(tert-butoxycarbonyl)amino]-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol):
426.47
Molecular Formula:
C23H26N2O6
InChi Key:
MVWPBNQGEGBGRF-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Boc-D-Dap Fmoc , Advanced ChemTech
CAS: 131570-56-4 Molecular Formula: C23H26N2O6 Molecular Weight (g/mol): 426.47 InChI Key: MVWPBNQGEGBGRF-UHFFFAOYNA-N IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid SMILES: CC(C)(C)OC(=O)NC(CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O