accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131651-65-5

IUPAC Name: lithium(1+) nonafluorobutane-1-sulfonate
Synonyms: lithium(1+) nonafluorobutane-1-sulfonate
Molecular Weight (g/mol): 306.03
Molecular Formula: C4F9LiO3S
InChi Key: FEDFHMISXKDOJI-UHFFFAOYSA-M
SMILES: [Li+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results