Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Nonafluoro-1-butanesulfonate 95.0+%, TCI America™
SDP

Catalog No. N071025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N071025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N071025G Supplier TCI America Supplier No. N071025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 131651-65-5
Molecular Formula C4F9LiO3S
Molecular Weight (g/mol) 306.027
MDL Number MFCD03094160
InChI Key FEDFHMISXKDOJI-UHFFFAOYSA-M
Synonym Lithium Perfluoro-1-butanesulfonate, Nonafluoro-1-butanesulfonic Acid Lithium Salt, Perfluoro-1-butanesulfonic Acid Lithium Salt
PubChem CID 23681910
IUPAC Name lithium;1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonate
SMILES [Li+].C(C(C(F)(F)S(=O)(=O)[O-])(F)F)(C(F)(F)F)(F)F

Specifications

Color White
Quantity 25 g
Formula Weight 306.03
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lithium Nonafluoro-1-butanesulfonate
EINECSNumber (2)-2810
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.