accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOOH(E/Z)

CAS RN 131674-41-4

IUPAC Name: 3-(2-methoxypyridin-3-yl)prop-2-enoic acid
Synonyms: 3-(2-methoxypyridin-3-yl)prop-2-enoic acid
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: MTVRDAQJMCORAQ-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(O)=O)C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results