CAS RN 1317-65-3
IUPAC Name:
calcium carbonate
Synonyms:
calcium carbonate calciumcarbonat carbonato de calcio carbonic acid calcium salt (1:1) CaCO3 kohlensaurer kalk carbonate de calcium calcite kalziumkarbonat calcite powder
Molecular Weight (g/mol):
100.09
Molecular Formula:
CCaO3
InChi Key:
VTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES:
[Ca++].[O-]C([O-])=O
1
–
2
of
2
results
1
Calcium Carbonate Marble Chips, Lab Grade, Innovating Science™
CAS: 1317-65-3 Molecular Weight (g/mol): 100.09 g/mol
2
Calcium Carbonate Natural Chalk, Lab Grade, Innovating Science™
CAS: 1317-65-3 Molecular Weight (g/mol): 100.09 g/mol