Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Carbonate Marble Chips, Lab Grade, Innovating Science™
SCI_ED

Catalog No. S000578 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000578 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning: May cause damage to organs (lungs) through prolonged or repeated exposure (inhalation). WARNING! This product can expose you to a chemical, crystalline silica, which is known to the State of California to cause cancer.
Catalog No. S000578 Supplier Aldon Corporation Supplier No. IS12024
Add to Cart
Add to Cart
Warning: May cause damage to organs (lungs) through prolonged or repeated exposure (inhalation). WARNING! This product can expose you to a chemical, crystalline silica, which is known to the State of California to cause cancer.

Chemical Identifiers

CAS 1317-65-3 , 7732-18-5
Molecular Weight (g/mol) 100.09 g/mol

Specifications

Physical Form Flakes or Powder
Quantity 500 g
Percent Purity > 99.5%
Grade Laboratory
Packaging HDPE
Chemical Name or Material Calcium Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.