accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHHN

CAS RN 13170-62-2

IUPAC Name: N-({4-[(phenylamino)methyl]phenyl}methyl)aniline
Synonyms: N-({4-[(phenylamino)methyl]phenyl}methyl)aniline
Molecular Weight (g/mol): 288.39
Molecular Formula: C20H20N2
InChi Key: DXWQPWMYKQYRDS-UHFFFAOYSA-N
SMILES: C(NC1=CC=CC=C1)C1=CC=C(CNC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results