Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha'-Dianilino-p-xylene 98.0+%, TCI America™
SDP

Catalog No. D21435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21435G 5 g
D21431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21435G Supplier TCI America Supplier No. D21435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13170-62-2
Molecular Formula C20H20N2
Molecular Weight (g/mol) 288.394
MDL Number MFCD00143578
InChI Key DXWQPWMYKQYRDS-UHFFFAOYSA-N
PubChem CID 607368
IUPAC Name N-[[4-(anilinomethyl)phenyl]methyl]aniline
SMILES C1=CC=C(C=C1)NCC2=CC=C(C=C2)CNC3=CC=CC=C3

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 5 g
Formula Weight 288.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha′-Dianilino-p-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.