CAS RN 13171-18-1
CAS RN 13171-18-1
IUPAC Name:
1,1,1,3,3,3-hexafluoro-2-methoxypropane
Synonyms:
1,1,1,3,3,3-hexafluoro-2-methoxypropane
Molecular Weight (g/mol):
182.07
Molecular Formula:
C4H4F6O
InChi Key:
VNXYDFNVQBICRO-UHFFFAOYSA-N
SMILES:
COC(C(F)(F)F)C(F)(F)F
1
–
1
of
1
results
1
Hexafluoroisopropyl Methyl Ether 98.0+%, TCI America™
CAS: 13171-18-1 Molecular Formula: C4H4F6O Molecular Weight (g/mol): 182.065 MDL Number: MFCD00221772 InChI Key: VNXYDFNVQBICRO-UHFFFAOYSA-N Synonym: 1,1,1,3,3,3-Hexafluoro-2-methoxypropane, Isoindoklon PubChem CID: 25749 IUPAC Name: 1,1,1,3,3,3-hexafluoro-2-methoxypropane SMILES: COC(C(F)(F)F)C(F)(F)F