accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3C(E/Z)

CAS RN 13172-91-3

IUPAC Name: 5-methylhept-3-ene
Synonyms: 5-methylhept-3-ene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: YMNTZRCUPAYGLG-UHFFFAOYNA-N
SMILES: CCC=CC(C)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results