Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-3-heptene (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. M09935ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09935ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M09935ML Supplier TCI America Supplier No. M09935ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13172-91-3
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00049124
InChI Key YMNTZRCUPAYGLG-UHFFFAOYSA-N
PubChem CID 143054
IUPAC Name 5-methylhept-3-ene
SMILES CCC=CC(C)CC

Specifications

Color Yellow
Quantity 5 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Methyl-3-heptene (cis- and trans- mixture)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.