accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131747-36-9

H3COONN

CAS RN 131747-36-9

IUPAC Name: (2-cyanopyridin-3-yl)methyl acetate
Synonyms: (2-cyanopyridin-3-yl)methyl acetate
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: HFTYCPJXTBOSFA-UHFFFAOYSA-N
SMILES: CC(=O)OCC1=C(N=CC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results