accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3SiCH3H3CCH3

CAS RN 13183-70-5

IUPAC Name: trimethyl[4-(trimethylsilyl)phenyl]silane
Synonyms: trimethyl[4-(trimethylsilyl)phenyl]silane
Molecular Weight (g/mol): 222.48
Molecular Formula: C12H22Si2
InChi Key: NVRBTKMAZQNKPX-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CC=C(C=C1)[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results