CAS RN 131848-97-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Ro-31-8425
A potent, cell-permeable, reversible, ATP-competitive, and selective protein kinase C inhibitor (IC50 = 15 nM for rat brain PKC).