accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131860-33-8

IUPAC Name: methyl (2E)-2-(2-{[6-(2-cyanophenoxy)pyrimidin-4-yl]oxy}phenyl)-3-methoxyprop-2-enoate
Synonyms: azoxystrobin
Molecular Weight (g/mol): 403.39
Molecular Formula: C22H17N3O5
InChi Key: WFDXOXNFNRHQEC-GHRIWEEISA-N
SMILES: CO\C=C(\C(=O)OC)C1=CC=CC=C1OC1=CC(OC2=CC=CC=C2C#N)=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results