accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHNN

CAS RN 131878-23-4

IUPAC Name: tert-butyl N-(1-benzylpyrrolidin-3-yl)carbamate
Synonyms: tert-butyl N-(1-benzylpyrrolidin-3-yl)carbamate
Molecular Weight (g/mol): 276.38
Molecular Formula: C16H24N2O2
InChi Key: PHOIDJGLYWEUEK-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1CCN(CC2=CC=CC=C2)C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results