accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1319-46-6

IUPAC Name: tris(λ²-lead(2+)) dihydroxide dicarbonate
Synonyms: white lead
Molecular Weight (g/mol): 775.6
Molecular Formula: C2H2O8Pb3
InChi Key: RYZCLUQMCYZBJQ-UHFFFAOYSA-H
SMILES: [OH-].[OH-].[Pb++].[Pb++].[Pb++].[O-]C([O-])=O.[O-]C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (4)
  • (7)
  • (4)
  • (2)