Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(Ii) Carbonate Basic, Solstice

Catalog No. 60044701 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
50 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-701 6 x 1 kg
60-019-07 1 kg
60-008-92 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-044-701 Supplier Solstice Supplier No. 115136X1KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss., white lead,≥77% Pb basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1319-46-6
MDL Number MFCD00078155

Specifications

Quantity 6 x 1 kg
Linear Formula (PbCO3)2·Pb(OH)2
UN Number UN3077
Formula Weight 775.63
Percent Purity ≥77% (Pb basis)
Grade puriss.
Chemical Name or Material Lead(Ii) Carbonate Basic
Hazard Statement P201-P260-P280-P301 + P312 + P330-P308 + P313
Precautionary Statement H302 + H332-H360Df-H373-H410
DOTInformation 9 / PGIII
EINECSNumber 215-290-6
RTECSNumber OF9275000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.