accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13194-69-9

IUPAC Name: 2-iodo-5-methylaniline
Synonyms: 2-iodo-5-methylaniline
Molecular Weight (g/mol): 233.05
Molecular Formula: C7H8IN
InChi Key: KXPBTNCFONSVIA-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C(I)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results