Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodo-5-methylaniline 98.0+%, TCI America™
SDP

Catalog No. I10345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10345G 5 g
I10341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10345G Supplier TCI America Supplier No. I10345G
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13194-69-9
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.052
MDL Number MFCD00833395
InChI Key KXPBTNCFONSVIA-UHFFFAOYSA-N
Synonym 3-Amino-4-iodotoluene
PubChem CID 2736659
IUPAC Name 2-iodo-5-methylaniline
SMILES CC1=CC(=C(C=C1)I)N

Specifications

Melting Point 36°C
Quantity 5 g
Formula Weight 233.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodo-5-methylaniline
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.