accessibility menu, dialog, popup

UserName

Viewing profile as user:
MnMnOHOHOPOOOOHOHOPOOO

CAS RN 1320-46-3

IUPAC Name: dimanganese(2+) 3-(phosphonatooxy)pentane-1,5-diol 3-(phosphonatooxy)propane-1,2-diol
Synonyms: dimanganese(2+) 3-(phosphonatooxy)pentane-1,5-diol DL-glycerol 3-phosphate
Molecular Weight (g/mol): 478.04
Molecular Formula: C8H18Mn2O12P2
InChi Key: PNNHLULHRSZWPG-UHFFFAOYNA-J
SMILES: [Mn++].[Mn++].OCC(O)COP([O-])([O-])=O.OCCC(CCO)OP([O-])([O-])=O
Grade 
  • (4)
Percent Purity 
  • (4)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)