Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Glycerophosphate, Powder, FCC, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18604203 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-203 50 kg Poly Bottle
18-604-200 500 g Poly Bottle
18-604-201 2.5 kg Poly Bottle
18-604-202 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-203 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. MA16350KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

MA163, 1320-46-3, C3H7MnO6P·xH2O

Spectrum™ Chemical Manganese Glycerophosphate, Powder, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1320-46-3
Molecular Formula C8H18Mn2O12P2
Molecular Weight (g/mol) 478.04
InChI Key PNNHLULHRSZWPG-UHFFFAOYNA-J
IUPAC Name dimanganese(2+) 3-(phosphonatooxy)pentane-1,5-diol 3-(phosphonatooxy)propane-1,2-diol
SMILES [Mn++].[Mn++].OCC(O)COP([O-])([O-])=O.OCCC(CCO)OP([O-])([O-])=O

Specifications

CAS Max % 100%
Physical Form Powder, Needles or Crystals
Quantity 50 kg
Percent Purity 98 to 100.5%
Grade FCC
Loss on Drying 12%
Packaging Poly Bottle
Chemical Name or Material Manganese Glycerophosphate, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.