accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOBrBrBrBr

CAS RN 13209-16-0

IUPAC Name: 1,2-bis(dibromomethyl)-4-nitrobenzene
Synonyms: 1,2-bis(dibromomethyl)-4-nitrobenzene
Molecular Weight (g/mol): 466.75
Molecular Formula: C8H5Br4NO2
InChi Key: KYHYXMVGEXICQQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(C(Br)Br)=C(C=C1)C(Br)Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results