Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-alpha,alpha,alpha',alpha'-tetrabromo-o-xylene 95.0+%, TCI America™
SDP

Catalog No. N05345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N05345G Supplier TCI America Supplier No. N05345G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13209-16-0
Molecular Formula C8H5Br4NO2
Molecular Weight (g/mol) 466.75
MDL Number MFCD00143350
InChI Key KYHYXMVGEXICQQ-UHFFFAOYSA-N
Synonym 3,4-Bis(dibromomethyl)nitrobenzene
PubChem CID 633527
IUPAC Name 1,2-bis(dibromomethyl)-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC(C(Br)Br)=C(C=C1)C(Br)Br

Specifications

Melting Point 124°C
Color Yellow
Quantity 5 g
UN Number 1759
Formula Weight 466.75
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-alpha,alpha,alpha′,alpha′-tetrabromo-o-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.