accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CCH2

CAS RN 1321-74-0

IUPAC Name: 1,2-diethenylbenzene
Synonyms: divinylbenzene
Molecular Weight (g/mol): 130.19
Molecular Formula: C10H10
InChi Key: MYRTYDVEIRVNKP-UHFFFAOYSA-N
SMILES: C=CC1=CC=CC=C1C=C
Percent Purity 
  • (2)
  • (5)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (5)
  • (2)
  • (5)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (7)
  • (6)
  • (1)