Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Divinylbenzene

Catalog No. 414565250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-456-5250ML 250 mL
41-456-51L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-456-5250ML Supplier Sigma Aldrich Supplier No. 414565250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Divinylbenzene (DVB) is a bis unsaturated monomer. DVB along with styrene as a co monomer is used in producing cross linked polymers. Several examples have reported. Polymerization with high concentrations of DVB produces popcorn polymers.DVB is used in polymeric core shell nanoparticles and as a crosslinking agent. DVB may be used as a monomer for the synthesis of highly crosslinked macroporous styrene/DVB polymers. It may also be used to synthesize monodispersed poly-DVB microspheres.

Specifications

CAS 1321-74-0
Density 0.914 g/mL (at 25°C)
Boiling Point 195°C
Molecular Formula C10H10
Refractive Index n20/D 1.561 (literature)
Linear Formula C6H4(CH=CH2)2
Quantity 250 mL
MDL Number MFCD01778656
Synonym Diethenylbenzene
Molecular Weight (g/mol) 130.19
Percent Purity 80%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.