accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FFF

CAS RN 132123-39-8

IUPAC Name: 3,4,5-trifluoro-4'-(4-propylcyclohexyl)-1,1'-biphenyl
Synonyms: 3,4,5-trifluoro-4'-(4-propylcyclohexyl)-1,1'-biphenyl
Molecular Weight (g/mol): 332.41
Molecular Formula: C21H23F3
InChi Key: RRKRBRVTKIRLHH-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results