accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 132196-28-2

IUPAC Name: (2-{[3-(hexadecyloxy)-2-(icosa-5,8,11,14,17-pentaenoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Synonyms: (2-{[3-(hexadecyloxy)-2-(icosa-5,8,11,14,17-pentaenoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Molecular Weight (g/mol): 766.1
Molecular Formula: C44H80NO7P
InChi Key: YHSSHPWVFALMBA-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCOCC(COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCC=CCC=CCC=CCC=CCC=CCC
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results