accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13224-94-7

IUPAC Name: (2S,3R,4S,5R,6R)-6-(hydroxymethyl)-4-methoxyoxane-2,3,5-triol
Synonyms: (2S,3R,4S,5R,6R)-6-(hydroxymethyl)-4-methoxyoxane-2,3,5-triol
Molecular Weight (g/mol): 194.18
Molecular Formula: C7H14O6
InChi Key: SCBBSJMAPKXHAH-OVHBTUCOSA-N
SMILES: CO[C@@H]1[C@@H](O)[C@@H](O)O[C@H](CO)[C@H]1O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results