Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-O-methyl-D-glucose, 99.1%, MP Biomedicals™

Catalog No. ICN15165110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15165110 10 g
ICN15165101 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15165110 Supplier MP Biomedicals, Inc Supplier No. 0215165110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13224-94-7
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063266
InChI Key SCBBSJMAPKXHAH-OVHBTUCOSA-N
Synonym 3-o-methyl-alpha-d-glucopyranose, 3-o-methyl-a-d-glucopyranose, 2s,3r,4s,5r,6r-6-hydroxymethyl-4-methoxytetrahydro-2h-pyran-2,3,5-triol, 2s,3r,4s,5r,6r-6-hydroxymethyl-4-methoxyoxane-2,3,5-triol, 3-o-methyl-, a-d-glucopyranose, 3-o-methyl-alpha-d-glucose, a-d-glucopyranose,3-o-methyl, alpha-d-glucopyranose, 3-o-methyl, 3-o-methyl-d-glucopyranose
PubChem CID 83246
IUPAC Name (2S,3R,4S,5R,6R)-6-(hydroxymethyl)-4-methoxyoxane-2,3,5-triol
SMILES CO[C@@H]1[C@@H](O)[C@@H](O)O[C@H](CO)[C@H]1O

Specifications

Color White
Quantity 10 g
Formula Weight 194.2
Percent Purity 0.991
Physical Form Powder
Chemical Name or Material 3-O-methyl-D-glucose
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.