accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CN

CAS RN 13228-39-2

IUPAC Name: 1-ethyl-2-phenyl-1H-indole
Synonyms: 1-ethyl-2-phenylindole
Molecular Weight (g/mol): 221.3
Molecular Formula: C16H15N
InChi Key: RAKSXVONTIQCGY-UHFFFAOYSA-N
SMILES: CCN1C(=CC2=CC=CC=C12)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)