Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-2-phenylindole, 98%, Thermo Scientific™

Catalog No. AAL1122818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1122818 50 g
AAL1122809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1122818 Supplier Thermo Scientific Chemicals Supplier No. L1122818

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13228-39-2
Molecular Formula C16H15N
Molecular Weight (g/mol) 221.303
MDL Number MFCD00031481
InChI Key RAKSXVONTIQCGY-UHFFFAOYSA-N
Synonym 1-ethyl-2-phenyl-1h-indole, n-ethyl-2-phenylindole, pubchem7337, 2-phenyl-1-ethylindole, acmc-1chnt, n-ethyl 2-phenyl indole, 1h-indole,1-ethyl-2-phenyl, 1h-indole, 1-ethyl-2-phenyl
PubChem CID 83248
IUPAC Name 1-ethyl-2-phenylindole
SMILES CCN1C2=CC=CC=C2C=C1C3=CC=CC=C3

Specifications

Melting Point 85°C to 86°C
Quantity 50 g
Beilstein 168581
Formula Weight 221.3
Percent Purity 98%
Chemical Name or Material 1-Ethyl-2-phenylindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.