accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1323-03-1

H3COOCH3OH

CAS RN 1323-03-1

IUPAC Name: tetradecyl 2-hydroxypropanoate
Synonyms: myristyl lactate
Molecular Weight (g/mol): 286.46
Molecular Formula: C17H34O3
InChi Key: BORJONZPSTVSFP-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCOC(=O)C(C)O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results