Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecyl Lactate 75.0+%, TCI America™
SDP

Catalog No. T223125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T223125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T223125G Supplier TCI America Supplier No. T223125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1323-03-1
Molecular Formula C17H34O3
Molecular Weight (g/mol) 286.456
InChI Key BORJONZPSTVSFP-UHFFFAOYSA-N
Synonym Lactic Acid Tetradecyl Ester, Myristyl Lactate, Tetradecyl 2-Hydroxypropionate
PubChem CID 64676
IUPAC Name tetradecyl 2-hydroxypropanoate
SMILES CCCCCCCCCCCCCCOC(=O)C(C)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 286.46
Percent Purity ≥75.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Tetradecyl Lactate
EINECSNumber (2)-1374
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.