CAS RN 132307-50-7
CAS RN 132307-50-7
IUPAC Name:
2,5-dioxopyrrolidin-1-yl (2S)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoate
Synonyms:
2,5-dioxopyrrolidin-1-yl (2S)-6-[(tert-butoxycarbonyl)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoate
Molecular Weight (g/mol):
565.62
Molecular Formula:
C30H35N3O8
InChi Key:
HONZVSWDWBWWMH-DEOSSOPVSA-N
SMILES:
CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(=O)ON1C(=O)CCC1=O
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Lys Boc -OSu, Advanced ChemTech
CAS: 132307-50-7 Molecular Formula: C30H35N3O8 Molecular Weight (g/mol): 565.62 MDL Number: MFCD00153359 InChI Key: HONZVSWDWBWWMH-DEOSSOPVSA-N IUPAC Name: 2,5-dioxopyrrolidin-1-yl (2S)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoate SMILES: CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(=O)ON1C(=O)CCC1=O