accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13231-81-7

IUPAC Name: 3-methylhexan-1-ol
Synonyms: 3-methylhexan-1-ol
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: YGZVAQICDGBHMD-UHFFFAOYNA-N
SMILES: CCCC(C)CCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results